Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ G-Tek™ Economy Grade Foam Nitrile Coated Seamless Knit Nylon Gloves

Catalog No. 191301175b Shop All Protective Industrial Products Inc. Products
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Seamless nylon shell increases comfort, finger dexterity, and breathability. The foam nitrile coating is designed with a cell structure to disperse fluids on contact for enhanced grip capabilities.

  • Knit wrist helps to prevent dirt and debris from entering the glove
  • Foam nitrile coating is breathable and durable
  • Gloves are resistant to chemicals, water, and ultraviolet light
  • Ideal for part handling, equipment maintenance, and assembly
  • Color coded hems for easy size identification

Specifications

Construction Seamless Knit
Coating Material Nitrile
Color Gray, White
Exterior Finish No Texture
Grade Economy
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size Small
For Use With (Application) Parts Handling, Assembly, and Equipment Maintenance
Glove Cuff Style Continuous Knit
Hem Color Red
Palm Type Foam Nitrile
Series 34-C234
Length (English) 8.9 in.
Length (Metric) 22.5 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.