Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIP™ Aramid Fiber Knit Wrist Sleeves
Shop All Protective Industrial Products Inc. Products
Click to view available options
:
Aramid/Cotton; Length: 56cm (22 in.); EN cut level: 2
Description
- Kevlar™ Aramid fibers knit wrist
- Blue/Gold elastic end
- Thumbhole
Ideal for applications where arm cut protections is needed, such as assembly, plastic molding, metal fabrication, and automotive work
Compliance
EN Cut levels 2 and 3
Specifications
Specifications
| Material | Aramid Fiber, Cotton |
| Cuff Style | Knit |
| Length (English) | 22 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction