Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Half-Finger Medium Weight Seamless Knit Cotton/Polyester Gloves

Catalog No. p-3347694 Shop All Protective Industrial Products Inc. Products
Click to view available options
Size:
Small
Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Seamless knit cotton/polyester shell is comfortable and flexible, while the half-finger design increases dexterity and sensitivity.

  • Knit wrist helps prevent dirt and debris from entering the glove
  • Reversible design allows gloves to be replaced one glove at a time, rather than in pairs
  • Half-finger design increases dexterity, sensitivity, and breathability
  • Complies with FDA food handling requirements 21 CFR, Part 177
  • Can be used alone, or as a glove liner

Specifications

Certifications/Compliance 21 CFR, 170-199
Construction Seamless Knit
Coating Material Uncoated
Color Natural
Material Polycotton
Product Type Seamless Knit Gloves
Size Small
Style Reversible
Finger Style Half
For Use With (Application) Warehousing, Parts Handling, Assembly, and Food Processing
Glove Cuff Style Continuous Knit
Series 35-C119
Yarn Gauge 7
Length (English) 6.5 in.
Length (Metric) 16.5 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.