Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Kut-Gard™ Aramid Fiber Gloves, Medium-Weight

Catalog No. 19100519 Shop All Protective Industrial Products Inc. Products
Click to view available options
Length (English):
9 in.
10 in.
10.5 in.
11 in.
Length (Metric):
22.9 cm
25.4 cm
26.7 cm
27.9 cm
Size:
Small
Medium
Large
X-Large
Glove Cuff Color:
Green
Red
White
Yellow
This item is not returnable. View return policy
This item is not returnable. View return policy

Aramid fiber gloves provide a comfortable and enduring protection against heat, cuts, lacerations and abrasions.

  • Aramid fiber provides high tensile strength relative to its weight and up to five times stronger than steel
  • Ideal for use in the automotive industry, light heat applications, glass operations and metal stamping
  • Seamless knit construction protects hands without sacrificing comfort or dexterity
  • Ambidextrous pattern
  • Can be used as gloves or glove liners
  • Color-coded cuff hems for easy size identification

Specifications

Construction Seamless Knit
Color Yellow
Cut Resistance Rating ANSI A2
Hand Ambidextrous
Material Aramid Fiber
Product Type Cut-resistant Gloves
Size X-Large
For Use With (Application) Automotive industry, light heat applications, glass operations and metal stamping
Glove Cuff Color Green
Glove Cuff Style Elasticated
Hem Color Green
Model 07-K300/XL
Series Kut-Gard™ 07-K300
Yarn Gauge 7
Length (English) 11 in.
Length (Metric) 27.9 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.