Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIP™ Kut-Gard™ PolyKor™ Seamless Knit Antimicrobial Gloves
Shop All Protective Industrial Products Inc. Products
Click to view available options
Length (English):
8 in.
8.5 in.
10 in.
10.5 in.
11 in.
Length (Metric):
20.3 cm
21.6 cm
25.4 cm
26.7 cm
27.9 cm
Size:
X-Small
Small
Medium
Large
X-Large
Glove Cuff Color:
Black
Blue
Green
White
Yellow
Description
- Seamless knit construction provides comfort, fit, and dexterity
- PolyKor™ engineered yarns offer exceptional cut and slash resistance
- Ambidextrous pattern permits glove to be worn on either hand, extending wear and reducing costs
- Special non-latex cuff dramatically reduces cuff blowout, even after multiple launderings in bleach
- Can be worn on its own or as a glove liner
- Good gripping properties
- Shrink resistant
- Made from component materials that comply with Federal Regulations for food contact 21 CFR, 170-199
- Antimicrobial gloves are non-toxic and non-corrosive and comply with FDA, EPA, NSF and USDA standards for food contact
- Products constructed with PolyKor engineered yarn are suitable for protection against mechanical hazards in the toughest environments
- PolyKor products are also highly resistant to abrasion and chemicals, so you can rely on them to provide long-lasting protection
Specifications
Specifications
Certifications/Compliance | FDA 21 CFR, 170-199 |
Construction | Seamless Knit |
Coating Material | Uncoated |
Color | White |
Cut Resistance Rating | ANSI A5 |
Material | Stainless Steel Fiber, Synthetic Fiber |
Product Type | Cut-resistant Gloves |
Size | Small |
For Use With (Application) | Application where sharp cutting instruments are used, such as food service, food processing and glass/metal handling |
Glove Cuff Color | Yellow |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction