Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Regular Grade Top Grain Cowhide Leather Drivers Gloves

Catalog No. 19121019C Shop All Protective Industrial Products Inc. Products
Click to view available options
Size:
X-Small
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Top grain cowhide leather construction provides comfort, durability, abrasion resistance, and breathability. Drivers gloves feature a slip-on cuff to enhance fit and comfort.

  • Top grain cowhide leather is smooth, provides durability and dexterity
  • Keystone thumb is ergonomic, reduces hand fatigue, offers dexterity and durability
  • Elastic closure sewn inside glove for safety and comfort
  • Cotton color binding denotes sizing
  • Meets the content requirements for Chromium VI free leather

Specifications

Color Natural
Grade Regular
Hand Specific
Material Top Grain Cowhide Leather
Product Type Driver Gloves
Size Large
For Use With (Application) Construction, Machine Operation, Light Duty Welding, Utility Work, Farming, and Roping
Glove Cuff Style Open
Hem Color Brown
Series 68-163
Thumb Style Keystone
Length (English) 9.8 in.
Length (Metric) 24.8 cm
Weight 33 lb. (Case)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.