Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ MaxiFlex™ Active™ Ultra Lightweight Nitrile Coated Nylon Lycra™ Gloves

Catalog No. 191501630 Shop All Protective Industrial Products Inc. Products
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

MicroFoam Nitrile coating is compatible with light oils to provide abrasion resistance, and enhanced grip capabilities. Coating is 25% thinner than most foam nitrile gloves, while offering twice the amount of mechanical performance.

  • Knit wrist prevents dirt and debris from entering the glove
  • Patented micro-foam nitrile coating offers 360° breathability
  • 25% thinner than most foam nitrile gloves on the market, while offering twice the mechanical performance
  • Glove fit mimics the hand at rest to reduce hand fatigue and increase comfort
  • Constructed using the latest knitting technology to produce smooth and rounded fingertips for increased sensitivity
  • Micro-cup finish allows for a controlled grip in light oil applications
  • Gloves are laundered prior to final packaging to ensure that they are clean and safe for use
  • All ingredients used in the manufacturing and construction of this product are compliant with REACH legislation
  • Oeko-Tex™ Association has evaluated and certified this product as skin safe from the second it comes into contact with the skin
  • Skin Health Alliance has awarded professional dermatological accreditation following review of the scientific dossier behind this product
  • Complies with FDA food handling requirements 21 CFR, Part 177
  • 100% Silicone free

Specifications

Construction Seamless Knit
Coating Material Nitrile Foam
Color Blue
Exterior Finish No Texture
Hand Specific
Material Nylon, Spandex
Product Type Seamless Knit Gloves
Size Large
Style Standard
EN388 Standard 4131
For Use With (Application) General Assembly, Micro-Engineering, Automotive Components, Horticulture, and Painting
Glove Cuff Style Continuous Knit
Series 34-824
Length (English) 8.7 in.
Length (Metric) 22 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.