Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP Mouse anti-Human, Clone-PIP/1571, Novus Biologicals™
SDP

Catalog No. NB254451100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
200 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB254451100 100 μg
NB254451200 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB254451100 Supplier Novus Biologicals Supplier No. NBP254451100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PIP Monoclonal specifically detects PIP in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Protein Array, CyTOF-ready.

Specifications

Antigen PIP
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
Classification Monoclonal
Clone PIP/1571
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, CyTOF-ready
Formulation PBS with No Preservative
Gene Alias GCDFP15prolactin-inducible protein, GCDFP-15SABP, GPIP4gp17, Gross cystic disease fluid protein 15, prolactin-induced proteinSecretory actin-binding protein
Gene Symbols PIP
Host Species Mouse
Immunogen Recombinant human GCDFP-15 protein fragment (aa 41-146) (exact sequence is proprietary)
Molecular Weight of Antigen 15 kDa
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5304
Target Species Human
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.