PIP™ NOVAX™ Electrical Safety Kits

Colorful, electrical protection products to increase safety

Manufacturer: Protective Industrial Products, Inc. 150SK00/11

 View more versions of this product

Catalog No. 19-168-2131

  Check Availability
Add to Cart

    Specifications

    Certifications ASTM D 120, NFPA 70E
    Includes 1 pair of ESP gloves, 1 pair of goatskin protector and 1 nylon glove bag
    For Use With (Application) power generation, industrial plant maintenance, utilities
    Length (English) 11 in.
    Length (Metric) 27.9cm
    Material Natural Rubber
    Size 11
    Color Black
    Classification 00
    Arc Flash Yes
    Palm Type Contoured

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit