Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIP™ NOVAX™ Class 0 Electrical Safety Gloves Kit
Shop All Protective Industrial Products Inc. Products
Click to view available options
Color:
Black
Orange
Size:
8
9
10
11
Description
- Includes one pair of natural rubber electrical insulating gloves, a pair of top grain goatskin leather glove protectors (148-1000), and an 11 in. nylon glove bag
- Class 0 insulating gloves will protect workers from up to 1000 V
- Meets NFPA 70E standards for use around arc flash hazards
Specifications
Specifications
Certifications/Compliance | ANSI/ASTM D120, Class 0, Type I, NFPA 70E |
Applications | Industrial |
Color | Orange |
Material | Natural Rubber, Top Grain Goatskin Leather |
Product Type | Electrical Insulating Gloves Kit |
Size | 9 |
For Use With (Application) | Electrical Utility Work, Industrial Plant Maintenance, and Power Generation |
Glove Cuff Style | Straight |
Includes | Natural Rubber Electrical Insulating Gloves (147-0-11), Top Grain Goatskin Leather Glove Protectors (148-1000), and a Nylon Glove Bag (148-2136) |
Series | 147-SK-0 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction