Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIP™ Medium Weight Nylon Half-Finger Clean Environment Gloves
Shop All Protective Industrial Products Inc. Products
Click to view available options
Size:
Small
Medium
Large
X-Large
Description
- Seamless nylon knit shell is comfortable and reduces hand fatigue
- Nylon yarns absorb perspiration to leave hands cool and comfortable
- Knit wrist helps prevent dirt and debris from entering the glove
- Reversible design allows gloves to be replaced one at a time, rather than in pairs
- Half-finger design enhances dexterity
- Color coded hems for easy size identification
- Ideal for use where low lint properties are desired
- Can be used as a glove liner
Specifications
Specifications
Construction | Seamless Knit |
Coating Material | Uncoated |
Color | White |
Disposable | Reusable |
Material | Nylon |
Size | X-Large |
Style | Reversible |
Finger Style | Half |
For Use With (Application) | Computer Manufacturing, Product Assembly/Finishing/Handling, and Photography Labs |
Glove Cuff Style | Knit |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction