Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ CleanTeam™ Medium Weight Nylon Clean Environment Gloves

Catalog No. p-4379669 Shop All Protective Industrial Products Inc. Products
Click to view available options
Size:
Small
Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Seamless knit nylon shell is comfortable and reduces hand fatigue. Gloves are thin enough to be worn as liners, and ideal for use where low lint properties are required.

  • Knit Wrist helps prevent dirt and debris from entering the glove
  • Reversible design allows gloves to be replaced one at a time, rather than pairs
  • Nylon yarns absorb perspiration and leave hands cool and comfortable
  • Color coded hems for easy size identification

Specifications

Construction Seamless Knit
Coating Material Uncoated
Color White
Disposable Limited-use
Hand Specific
Latex Free Yes
Material Nylon
Product Type Inspection Gloves
Size Large
Sterility Non-sterile
Style Reversible
For Use With (Application) Product Handling, Electronics Assembly, Photography Labs, and Computer Manufacturing
Glove Cuff Style Knit
Hem Color White
Series 40-750
Yarn Gauge 10
Length (English) 9.1 in.
Length (Metric) 23 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.