Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Seamless Knit Polyester Gloves with Double-Sided PVC Grip

Catalog No. p-3348665 Shop All Protective Industrial Products Inc. Products
Click to view available options
Size:
Small
Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Gloves feature a PVC coating on both sides in a honeycomb criss-cross pattern for enhanced grip capabilities.

  • Seamless knit construction is comfortable and flexible
  • Polyester shell dries fast in wet applications
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Double-sided PVC coating in a honeycomb criss-cross pattern enhances grip, as well as cut and abrasion resistance

Specifications

Construction Seamless Knit
Coating Material PVC
Color Orange
Material Polyester
Product Type Seamless Knit Gloves
Size Small
For Use With (Application) Fishing, Glass/Plywood/Lumber Handling
Glove Cuff Style Continuous Knit
Series 39-3013
Yarn Gauge 10
Length (English) 9.7 in.
Length (Metric) 24.5 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.