Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ PowerGrab™ Premium Cotton/Polyester Gloves with Latex Coated MicroSurface Grip

Catalog No. 19165736A Shop All Protective Industrial Products Inc. Products
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Latex MicroSurface grip is infused with thousands of tiny suction cup pockets which create a vacuum effect that dispenses fluids away on contact for enhanced grip capabilities.

  • Seamless knit cotton/polyester shell is comfortable and flexible
  • Latex MicroSurface grip dispenses fluids away on contact
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Ergonomic shaped finger pattern for enhanced user comfort

Specifications

Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Latex
Color Brown
Hand Specific
Material Polycotton
Product Type Seamless Knit Gloves
Size X-Small
EN388 Standard 3242X
For Use With (Application) Assembly, Construction, Inspection, Material Handling, and Landscaping
Glove Cuff Style Continuous Knit
Series 39-C1500
Yarn Gauge 10
Length (English) 9.2 in.
Length (Metric) 23.5 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.