Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Seamless Knit Cotton/Polyester Gloves with Black PVC Coating

Catalog No. p-3543470 Shop All Protective Industrial Products Inc. Products
Click to view available options
Size:
Small
Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Seamless knit construction is comfortable and flexible, while PVC coating on the palm enhances grip capabilities.

  • Seamless knit shell is comfortable and flexible
  • Knit wrist helps prevent dirt and debris from entering the glove
  • PVC coating on the palm enhances grip and provides resistance to cuts and abrasion

Specifications

Coated Area Palm
Construction Seamless Knit
Coating Material PVC
Color Black, Natural
Hand Specific
Material Polycotton
Product Type Seamless Knit Gloves
Size Small
For Use With (Application) Package Handling, Warehousing, General Assembly, and Construction
Glove Cuff Style Continuous Knit
Series 37-C110PC
Yarn Gauge 7
Length (English) 8.9 in.
Length (Metric) 22.5 cm
Show More Show Less

WARNING: Cancer - www.P65Warnings.ca.gov

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.