Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ CleanTeam™ Seamless Knit Nylon Clean Environment Gloves - 13 Gauge
GSA_VA

Catalog No. p-2364278 Shop All Protective Industrial Products Inc. Products
Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy

Catalog No. 19-121-216A

Supplier: PIP™ 40730/S

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Seamless knit nylon construction is lightweight and reduces hand fatigue, while also absorbing perspiration to leave hands cool and comfortable.

  • Seamless knit shell is comfortable and reduces hand fatigue
  • Nylon yarns absorb perspiration to leave hands cool and comfortable
  • Knit Wrist helps prevent dirt and debris from entering the glove
  • Reversible design allows gloves to be replaced one glove at a time, rather than pairs
  • Ideal for use where low lint properties are desired
  • Can be used as a glove liner
  • Color coded hems for easy size identification

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Coated Area Uncoated
Construction Seamless Knit
Coating Material Uncoated
Color White
Disposable Limited-use
Hand Ambidextrous
Latex Free Yes
Material Nylon
Product Type Inspection Gloves
Size Small
Sterility Non-sterile
For Use With (Application) Computer Manufacturing, Electronics Assembly, Photography Labs, and Product Handling
Glove Cuff Style Knit
Hem Color Green
Series 40-730
Yarn Gauge 13
Length (English) 7.6 in.
Length (Metric) 19.3 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.