Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP2 Antibody (PIP2 2C11), Novus Biologicals™
SDP

Catalog No. NBP276433 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP276433 0.1 mg
NB413788 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP276433 Supplier Novus Biologicals Supplier No. NBP276433
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 1 publication

PIP2 Monoclonal specifically detects PIP2 in Non-species specific samples. It is validated for Chromatin Immunoprecipitation, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Functional.

Specifications

Antigen PIP2
Applications ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Functional Assay, Dot Blot
Classification Monoclonal
Clone PIP2 2C11
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Functional, Dot Blot, Chromatin Immunoprecipitation (ChIP)
Formulation PBS with 0.02% Sodium Azide
Gene Alias 1,2-Diacyl-sn-glycero-3-phospho-(1-D-myo-inositol 4,5-bisphosphate), Phosphatidylinositol 4,5-bisphosphate, Phosphatidylinositol biphosphate, Phosphatidylinositol diphosphate, PIP 2, PtdIns(4,5)P2
Host Species Mouse
Immunogen Liposomes (prepared in PBS with lipid A, phosphatidylcholine (PC) and cholesterol) containing synthetic di-palmitoyl PtdIns(4,5)P2.
Purification Method Thiophilic Chromatography
Quantity 0.1 mg
Primary or Secondary Primary
Test Specificity Cross reacts with PtdIns(4,5)P2, Ptd(4)P and Ptd(3,4,5)P3.
Target Species Non-species specific
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.