Learn More
Description
Specifically detects PI4K a, type I in Human samples, and it is validated for ELISA, Immunohistochemistry (Paraffin), Western Blotting
Specifications
Specifications
| Antigen | PIP5K1 alpha |
| Applications | Western Blot, ELISA, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.75 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1ug/ml, ELISA 1:10000, Immunohistochemistry 5 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml |
| Formulation | 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 30% Glycerol with No Preservative |
| Gene Accession No. | Q99755 |
| Gene Alias | 68 kDa type I phosphatidylinositol-4-phosphate 5-kinase alpha, EC 2.7.1, EC 2.7.1.68, Phosphatidylinositol-4-phosphate 5-kinase type I alpha, phosphatidylinositol-4-phosphate 5-kinase type-1 alpha, phosphatidylinositol-4-phosphate 5-kinase, type I, alpha, PIP5K1-alpha, PIP5KIalpha, PtdIns(4)P-5-kinase 1 alpha |
| Gene Symbols | PIP5K1A |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
