Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIP5K1C Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | PIP5K1C |
---|---|
Concentration | 0.3 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-5725
![]() ![]() |
Proteintech Group Inc
276401AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Phosphatidylinositol-4-phosphate-5-kinase (PIPK) synthesizes phosphatidylinositol-4,5-bisphosphate, which regulates various processes including cell proliferation, survival, membrane trafficking and cytoskeletal organization. The PIPK family is divided into type I, type II and type III. Each type of the PIPK family phosphorylates distinct substrates. They contain an activation loop, which determines their enzymatic specificity and subcellular targeting. The phosphatidylinositol-4-phosphate-5-kinase type I consists of three members, PIPK I alpha, beta and gamma, which are characterized by phosphorylating PI4P on the 5-hydroxyl. PIPK I alpha, designated PIPK I beta in mouse, is expressed in brain tissue. PIPK I beta, designated PIPK I alpha in mouse, is also called STM7. PIPK I gamma has two variants produced by alternative splicing which are expressed in lung, brain and kidneys.Specifications
PIP5K1C | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
O60331 | |
23396 | |
PIP5K1C Fusion Protein Ag26525 | |
Primary | |
-20°C | |
PIP5K1C |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
KIAA0589, LCCS3, PIP5K GAMMA, PIP5K1 gamma, PIP5K1C, PIP5Kgamma, PIP5KIgamma, PtdIns(4)P 5 kinase 1 gamma | |
PIP5K1C | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title