Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP5K2 alpha Antibody (OTI3D3), Novus Biologicals™
SDP

Catalog No. NBP203492 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP203492 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP203492 Supplier Novus Biologicals Supplier No. NBP203492
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PIP5K2 alpha Monoclonal specifically detects PIP5K2 alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen PIP5K2 alpha
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI3D3
Concentration 0.9 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:150
Gene Alias 1-phosphatidylinositol-4-phosphate kinase, 1-phosphatidylinositol-4-phosphate-5-kinase, 1-phosphatidylinositol-5-phosphate 4-kinase 2-alpha, Diphosphoinositide kinase 2-alpha, EC 2.7.1, EC 2.7.1.149, FLJ13267, phosphatidylinositol-4-phosphate 5-kinase, type II, alpha, Phosphatidylinositol-5-phosphate 4-kinase type II alpha, phosphatidylinositol-5-phosphate 4-kinase type-2 alpha, phosphatidylinositol-5-phosphate 4-kinase, type II, alpha, PI(5)P 4-kinase type II alpha, PIP4KII-alpha, PIP5K2, PIP5K2API5P4KA, PIP5KIIA, PIP5KIIalpha, PIP5KII-alpha, PIP5KIII, PIPK, PtdIns(4)P-5-kinase B isoform, PtdIns(4)P-5-kinase C isoform, PtdIns(5)P-4-kinase isoform 2-alpha, type II phosphatidylinositol-4-phosphate 5-kinase 53 K isoform
Gene Symbols PIP4K2A
Host Species Mouse
Immunogen Full length human recombinant protein of human PIP4K2A (NP_005019) produced in HEK293T cell.
Molecular Weight of Antigen 46 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5305
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.