Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP5K2 gamma Antibody, Novus Biologicals™
SDP

Catalog No. NBP219836 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP219836 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP219836 Supplier Novus Biologicals Supplier No. NBP219836
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PIP5K2 gamma Polyclonal specifically detects PIP5K2 gamma in Human samples. It is validated for Western Blot.

Specifications

Antigen PIP5K2 gamma
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q8TBX8
Gene Alias EC 2.7.1, EC 2.7.1.149, FLJ22055, phosphatidylinositol-4-phosphate 5-kinase, type II, gamma, Phosphatidylinositol-5-phosphate 4-kinase type II gamma, phosphatidylinositol-5-phosphate 4-kinase type-2 gamma, phosphatidylinositol-5-phosphate 4-kinase, type II, gamma, PI(5)P 4-kinase type II gamma, PIP4KII-gamma, PIP5K2C
Gene Symbols PIP4K2C
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human PIP5K2 gamma. The exact sequence is proprietary.
Molecular Weight of Antigen 47 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79837
Test Specificity Human. Rhesus Monkey (100%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.