Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Piperidine-1-sulfonyl chloride, 97%
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
Piperidine-1-sulfonyl chloride is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Piperidine-1-sulfonyl chloride is used as pharmaceutical intermediate.
Notes
Moisture sensitive. Incompatible with oxidizing agents.
Piperidine-1-sulfonyl chloride is used as pharmaceutical intermediate.
Notes
Moisture sensitive. Incompatible with oxidizing agents.
Specifications
Specifications
| Density | 1.308 |
| Refractive Index | 1.492 |
| Quantity | 5 g |
| UN Number | UN3265 |
| Sensitivity | Moisture sensitive |
| Formula Weight | 183.65 |
| Percent Purity | 97% |
| Chemical Name or Material | Piperidine-1-sulfonyl chloride |
Safety and Handling
| Hazard Category | H314-H335 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S. |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction