Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraethylene glycol dimethyl ether, 99%

Catalog No. AC174110025
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 143-24-8 | C10H22O5 | 222.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-24-8
Molecular Formula C10H22O5
Molecular Weight (g/mol) 222.28
MDL Number MFCD00008505
InChI Key ZUHZGEOKBKGPSW-UHFFFAOYSA-N
Synonym tetraglyme, tetraethylene glycol dimethyl ether, 2,5,8,11,14-pentaoxapentadecane, dimethoxytetraglycol, glyme 5, ansul ether 181at, dimethoxytetraethylene glycol, nissan uniox mm 200, bis 2-2-methoxyethoxy ethyl ether, methyltetraglyme200
PubChem CID 8925
ChEBI CHEBI:46785
IUPAC Name 1-methoxy-2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethane
SMILES COCCOCCOCCOCCOC

Specifications

Acidity 0.015% max.
Melting Point -30.0°C
Density 1.0090g/mL
Boiling Point 275.0°C to 276.0°C
Flash Point 141°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4310 to 1.4330
Linear Formula CH3(OCH2CH2)4OCH3
Quantity 2.5 kg
Beilstein 01,III,2107
Merck Index 15,9353
Specific Gravity 1.009
Solubility Information Solubility in water: soluble. Other solubilities: miscible with hydrocarbon solvents
Viscosity 4.1 mPa.s (20°C)
Formula Weight 222.28
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Tetraethylene glycol dimethyl ether
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May damage the unborn child.
Suspected of damaging fertility.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
EINECSNumber 205-594-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.