Learn More
Sartorius Pipet Charging Stand and Carousel
Shop All Sartorius Products
Click to view available options
Number of Channels:
1
4
Description
Includes
Universal AC-adaptor
For storing and charging Picus and eLINE mechanical and electronic pipets when not in use. Pipets should be stored in an upright position in order to avoid contamination from desks and benches.
Specifications
Specifications
For Use With | Pipette Tips |
Material | Polypropylene |
Number of Channels | 1 |
Type | Pipette Holder |
For Use With (Equipment) | Picus and eLINE Electronic Pipettes |
Includes | Universal AC adapter |