Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIR-A/B Antibody (404127), PE/Atto594, Novus Biologicals™
SDP

Catalog No. NB312075 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB312075 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB312075 Supplier Novus Biologicals Supplier No. FAB5308PEATT594
Add to Cart
Add to Cart

Rat Monoclonal Antibody

PIR-A/B Monoclonal specifically detects PIR-A/B in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen PIR-A/B
Applications Flow Cytometry
Classification Monoclonal
Clone 404127
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse PIR-A1, Gly24-Met650, Accession # A2NTJ8
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.