Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIR-B Antibody (326414), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB348776 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB348776 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB348776 Supplier Novus Biologicals Supplier No. FAB2754MFV610
Add to Cart
Add to Cart

Rat Monoclonal Antibody

PIR-B Monoclonal specifically detects PIR-B in Mouse samples. It is validated for CyTOF, Flow Cytometry, Western Blot.

Specifications

Antigen PIR-B
Applications CyTOF, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 326414
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD85a, HL9, ILT5, ILT-5, leukocyte immunoglobulin like receptor B3, LILRA6, LILRB3, LIR3, LIR-3, PIRB, PIR-B
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse PIR-B, Gly24-Gly635, Accession # AAH26937
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11025
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.