Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PISD Antibody (OTI4G5), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP273420A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP273420A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP273420A94 Supplier Novus Biologicals Supplier No. NBP273420AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PISD Monoclonal specifically detects PISD in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen PISD
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI4G5
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias DJ858B16, dJ858B16.2, DKFZp566G2246, EC 4.1.1.65, phosphatidylserine decarboxylase, phosphatidylserine decarboxylase proenzyme, PSD, PSDC, PSSC
Gene Symbols PISD
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 80-340 of human PISD (NP_055153) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 23761
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.