Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Pitavastatin calcium
GREENER_CHOICE

Catalog No. p-4817307
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

HMG-CoA reductase inhibitor

Synthetic inhibitor of HMG-CoA reductase (Ki = 1.7 nM). Lowers low-density lipoprotein (LDL) cholesterol levels. Displays minimal metabolism by cytochrome P450 enzymes, with little involvement of CYP3A4. Shown to improve cardiac function and survival in a rat model of hypertensive heart failure.

Chemical Identifiers

CAS 147526-32-7
Molecular Formula C50H46CaF2N2O8

Specifications

Quantity 10 mg
Formula Weight 440.5
Purity >99%
Molecular Weight (g/mol) 881.00
Chemical Name or Material Pitavastatin calcium
Recommended Storage Desiccate at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.