Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PITPN Antibody, Novus Biologicals™
SDP

Catalog No. NBP131042 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP131042 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP131042 Supplier Novus Biologicals Supplier No. NBP131042
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PITPN Polyclonal specifically detects PITPN in Human samples. It is validated for Western Blot.

Specifications

Antigen PITPN
Applications Western Blot
Classification Polyclonal
Concentration 1.0mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation PBS (pH 7.0) and 10% Glycerol with 0.01% Thimerosal
Gene Alias MGC99649, phosphatidylinositol transfer protein alpha isoform, phosphatidylinositol transfer protein, alpha, PI-TPalpha, PI-TP-alpha, PITPNphosphotidylinositol transfer protein, PtdIns transfer protein alpha, PtdInsTP alpha, VIB1A
Gene Symbols PITPNA
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human PITPN. The exact sequence is proprietary.
Molecular Weight of Antigen 32 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 5306
Test Specificity Tested in human. Predicted cross-reactivity based on sequence identity: Rhesus Monkey (100%). Not yet tested in any other species.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.