Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PITPN Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP198374
Description
PITPN Polyclonal specifically detects PITPN in Mouse samples. It is validated for Western Blot.Specifications
| PITPN | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| MGC99649, phosphatidylinositol transfer protein alpha isoform, phosphatidylinositol transfer protein, alpha, PI-TPalpha, PI-TP-alpha, PITPNphosphotidylinositol transfer protein, PtdIns transfer protein alpha, PtdInsTP alpha, VIB1A | |
| Rabbit | |
| 32 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_032876 | |
| PITPNA | |
| The immunogen for this antibody is PITPN - C-terminal region. Peptide sequence FTNFHRQLFCWLDKWVDLTMDDIRRMEEETKRQLDEMRQKDPVKGMTADD. | |
| Affinity purified | |
| RUO | |
| 5306 | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Rabbit, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction