Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PITPNB Antibody (OTI6H10) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP273421 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273421 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273421 Supplier Novus Biologicals Supplier No. NBP273421
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PITPNB Monoclonal specifically detects PITPNB in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen PITPNB
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI6H10
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:2000, Immunohistochemistry 1:150
Formulation Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
Gene Alias phosphatidylinositol transfer protein beta isoform, phosphatidylinositol transfer protein, beta, phosphotidylinositol transfer protein, beta, PI-TP-beta, PtdIns transfer protein beta, PtdInsTP, PtdInsTP beta, VIB1B
Gene Symbols PITPNB
Host Species Mouse
Immunogen Full length human recombinant protein of human PITPNB (NP_036531) produced in E.coli.
Purification Method Affinity Purified
Quantity 100 μg
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 23760
Reconstitution Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.