Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PITPNC1 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB329753 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB329753 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB329753 Supplier Novus Biologicals Supplier No. NBP297834PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PITPNC1 Polyclonal antibody specifically detects PITPNC1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen PITPNC1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias cytoplasmic phosphatidylinositol transfer protein 1, Mammalian rdgB homolog beta, M-rdgB beta, MrdgBbeta, phosphatidylinositol transfer protein, cytoplasmic 1, RDGBB, RDGBB1, rdgBbeta, RDGB-BETA, retinal degeneration B beta 1, Retinal degeneration B homolog beta
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human PITPNC1 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26207
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.