Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PITX1 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001546 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001546 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001546 Supplier Novus Biologicals Supplier No. NB1001546
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PITX1 Polyclonal specifically detects PITX1 in Human samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen PITX1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2500-1:10000, Immunoprecipitation 1-4 ug/mg of lysate
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. P78337
Gene Alias BFTpituitary homeobox 1, CCF, Hindlimb-expressed homeobox protein backfoot, Homeobox protein PITX1, paired-like homeodomain 1, pituitary otx-related factor, POTXpituitary homeo box 1, PTX1Paired-like homeodomain transcription factor 1hindlimb expressed homeobox protein backfoot
Gene Symbols PITX1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residues 50 and 100 of human Paired-Like Homeodomain Transcription Factor 1 using the numbering given in entry NP_002644.3 (GeneID 5307).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5307
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.