Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PITX2 Polyclonal Antibody, Active Motif™
SDP

Rabbit Polyclonal Antibody

$159.90 - $633.45

Specifications

Antigen PITX2
Concentration Lot-Specific
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-3320
SDP
Encompass
View Documents
Active Motif
61304
10 μL
Each for $159.90
Only null left
Add to Cart
 
50-199-3319
SDP
Encompass
View Documents
Active Motif
61303
100 μL
Each for $633.45
Only null left
Add to Cart
 
Description

Description

Pilx2 is a member of the RIEG/PITX homeobox family, which is in the bicoid class of homeodomain proteins. This protein acts as a transcription factor and regulates procollagen lysyl hydroxylase gene expression. It plays a role in the terminal differentiation of somatotroph and lactotroph cell phenotypes, is involved in the development of the eye, tooth and abdominal organs, and acts as a transcriptional regulator involved in basal and hormone-regulated activity of prolactin. Mutations in this protein are associated with Axenfeld-Rieger syndrome, iridogoniodysgenesis syndrome, and sporadic cases of Peters anomaly. A similar protein in other vertebrates is involved in the determination of left-right asymmetry during development.
Specifications

Specifications

PITX2
Western Blot
Unconjugated
Rabbit
Human
Q99697
5308
This PITX2 antibody was raised against a peptide within the N-terminal region of human PITX2.
Primary
-20°C, Avoid Freeze/Thaw Cycles
PITX2
Lot-Specific
Polyclonal
Liquid
RUO
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
9430085M16Rik; all1-responsive gene 1; ALL1-responsive protein ARP1; ARP1; BOS_6374; Brx1; BRX1 homeoprotein; Brx1b; Homeobox protein PITX2; homeodomain transcription factor 2; IDG2; IGDS; IGDS2; IHG2; IRID2; Munc30; Orthodenticle-like homeobox 2; Otlx2; paired like homeodomain 2; paired-like homeodomain 1; paired-like homeodomain 2; paired-like homeodomain transcription factor 1; Paired-like homeodomain transcription factor 2; Paired-like homeodomain transcription factor Munc 30; Pituitary homeobox 2; PITX1; PITX2; Pitx-2; Pitx2c; Ptx2; RGS; RIEG; RIEG bicoid-related homeobox transcription factor; rieg bicoid-related homeobox transcription factor 1; RIEG1; rPtx2; RS; S a 1 glycoprotein; S a1glycoprotein; S alpha 1 glycoprotein; S a 1 glycoprotein; S a1glycoprotein; Solurshin
PITX2
IgG
Antigen affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.