Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIWIL2 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB344046 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB344046 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB344046 Supplier Novus Biologicals Supplier No. NBP241126MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PIWIL2 Polyclonal antibody specifically detects PIWIL2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen PIWIL2
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Cancer/testis antigen 80, CT80mili, FLJ10351, HILIMiwi like, MGC133049, Mili, PIWIL1L, piwil2-like protein, piwi-like 2 (Drosophila), piwi-like protein 2
Host Species Rabbit
Immunogen Antibody was raised against a 14 amino acid synthetic peptide near the center of human PIWI-L2. The immunogen is located within amino acids 460 - 510 of PIWI-L2. Amino Acid Squence: LIHRPSERQDNHGM
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 55124
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.