Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIWIL2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224590SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224590SS 0.025 mg
NBP224590 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224590SS Supplier Novus Biologicals Supplier No. NBP224590SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PIWIL2 Polyclonal specifically detects PIWIL2 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen PIWIL2
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2-4 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. NP_067283.1
Gene Alias Cancer/testis antigen 80, CT80mili, FLJ10351, HILIMiwi like, MGC133049, Mili, PIWIL1L, piwil2-like protein, piwi-like 2 (Drosophila), piwi-like protein 2
Gene Symbols PIWIL2
Host Species Rabbit
Immunogen A portion of amino acids 30 to 80 of mouse Mili was used as immunogen.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55124
Test Specificity The amino acid sequence used as immunogen is 100% homologous in mouse and rat, 87% homologous in human and dog, and 81% homologous in opossum.
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.