Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIWIL4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP219845 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP219845 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP219845 Supplier Novus Biologicals Supplier No. NBP219845
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PIWIL4 Polyclonal specifically detects PIWIL4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen PIWIL4
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation PBS, 20% Glycerol (pH7) with 0.025% Proclin 300
Gene Accession No. Q7Z3Z4
Gene Alias DKFZp686P01248, FLJ36156, HIWI2MIWI2, Miwi2, PIWI, piwi-like 4 (Drosophila), piwi-like protein 4
Gene Symbols PIWIL4
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human PIWIL4. The exact sequence is proprietary.
Molecular Weight of Antigen 97 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 143689
Test Specificity Human. Species reactivity based on sequence identity: Monkey (98%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.