Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ PK-THPP

missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
50 mg
Description
PK-THPP is a potent TASK-3 channel blocker (IC50 = 35 and 300 nM for TASK-3 and TASK-1 respectively). Increases breathing rate and induces respiratory alkalosis in rats.
Specifications
Specifications
Quantity | 50 mg |
Product Type | PK-THPP |
Purity | >99% |
Molecular Weight (g/mol) | 468.59 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction