Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKA C alpha Antibody (6D2.1), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB002207 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002207 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002207 Supplier Novus Biologicals Supplier No. NBP262195FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PKA C alpha Monoclonal specifically detects PKA C alpha in Human samples. It is validated for Western Blot.

Specifications

Antigen PKA C alpha
Applications Western Blot
Classification Monoclonal
Clone 6D2.1
Conjugate DyLight 680
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias cAMP-dependent protein kinase catalytic subunit alpha, EC 2.7.11, EC 2.7.11.11, MGC102831, MGC48865, PKA C-alpha, PKACA, protein kinase A catalytic subunit, protein kinase, cAMP-dependent, catalytic, alpha
Gene Symbols PRKACA
Host Species Mouse
Immunogen Peptide corresponding to amino acids ESPAQNTAHLDQFERIK of human proteinkinase A c alpha (PKAc alpha).
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Phospho Specific, Protein Kinase, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5566
Test Specificity The antibody C-50 reacts with Cytokeratin peptides 5 and 18 (58 and 45 kDa). Cytokeratins are a member of intermediate filaments subfamily represented in epithelial tissues.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.