Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKA C beta Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB092824 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB092824 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB092824 Supplier Novus Biologicals Supplier No. NBP30346520UL

Rabbit Polyclonal Antibody

PKA C beta Polyclonal specifically detects PKA C beta in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.

Specifications

Antigen PKA C beta
Applications Western Blot, Immunohistochemistry, Immunofluorescence, KnockDown
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias cAMP-dependent protein kinase catalytic beta subunit isoform 4ab, cAMP-dependent protein kinase catalytic subunit beta, DKFZp781I2452, EC 2.7.11, EC 2.7.11.11, MGC41879, MGC9320, PKA C-beta, PKACb, protein kinase A catalytic subunit beta, protein kinase, cAMP-dependent, catalytic, beta
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-351 of human PKA C beta (NP_002722.1).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Phospho Specific, Protein Kinase, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5567
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.