Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKA C-gamma Antibody (2E4), Novus Biologicals™
SDP

Catalog No. NBP237487 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP237487 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP237487 Supplier Novus Biologicals Supplier No. NBP237487
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PKA C-gamma Monoclonal specifically detects PKA C-gamma in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen PKA C-gamma
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 20000
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000
Formulation Ascites with 0.03% Sodium Azide
Gene Alias cAMP-dependent protein kinase catalytic subunit gamma, EC 2.7.11, EC 2.7.11.11, KAPG, PKA C-gamma, PKACg, protein kinase, cAMP-dependent, catalytic, gamma, serine(threonine) protein kinase
Gene Symbols PRKACG
Host Species Mouse
Immunogen Purified recombinant fragment of human PRKACG expressed in E. Coli.
Molecular Weight of Antigen 40.4 kDa
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5568
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.