Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKC alpha Antibody (133), Biotin, Novus Biologicals™
SDP

Catalog No. NB382620 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB382620 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB382620 Supplier Novus Biologicals Supplier No. NBP308229B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PKC alpha Monoclonal antibody specifically detects PKC alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)

Specifications

Antigen PKC alpha
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 133
Conjugate Biotin
Formulation PBS
Gene Alias AAG6, aging-associated gene 6, EC 2.7.11, EC 2.7.11.13, MGC129901, PKC-A, PKC-alpha, PKCAMGC129900, PRKACA, protein kinase C alpha type, protein kinase C, alpha
Host Species Mouse
Immunogen Full-length protein corresponding to PKC alpha. Recognizes sequence PQFVHPILQSAV at the C terminus at PKCalpha. (Uniprot: P17252)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, mTOR Pathway, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5578
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.