Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKC alpha Antibody (MC5), HRP, Novus Biologicals™
SDP

Catalog No. NB600201H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600201H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600201H Supplier Novus Biologicals Supplier No. NB600201H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PKC alpha Monoclonal specifically detects PKC alpha in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Fish, Rabbit, Sheep, Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen PKC alpha
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone MC5
Conjugate HRP
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation PBS with No Preservative
Gene Alias AAG6, aging-associated gene 6, EC 2.7.11, EC 2.7.11.13, MGC129901, PKC-A, PKC-alpha, PKCAMGC129900, PRKACA, protein kinase C alpha type, protein kinase C, alpha
Gene Symbols PRKCA
Host Species Mouse
Immunogen Purified bovine brain Protein Kinase C
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cancer, Cellular Markers, mTOR Pathway, Phospho Specific, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5578
Test Specificity This antibody is capable of detecting PKC beta as well.
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Chicken, Fish, Rabbit, Sheep
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a Kappa
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.