Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKC epsilon Antibody, Novus Biologicals™
SDP

Catalog No. NB239447 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 ul
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB239447 25 μg
NB362532 100 ul
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB239447 Supplier Novus Biologicals Supplier No. NBP32122425UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PKC epsilon Polyclonal antibody specifically detects PKC epsilon in Human samples. It is validated for Immunofluorescence

Specifications

Antigen PKC epsilon
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias EC 2.7.11, EC 2.7.11.13, MGC125656, nPKC-epsilon, PKCEMGC125657, protein kinase C epsilon type, protein kinase C, epsilon
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: VDARGIAKVLADLGVTPDKITNSGQRRKKLIAGAESPQPASGSSPSEEDRSKSAPTSPCDQEIKELENNIRKALSFDNRGEEHRAASSPDGQLMSPGENGEVRQGQAKRLGLDEFNFIKVLG
Purification Method Affinity purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5581
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.