Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKC gamma Rabbit anti-Human, Mouse, Rat, Clone: JG69-32, Novus Biologicals™
SDP

Catalog No. NBP275621 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP275621 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP275621 Supplier Novus Biologicals Supplier No. NBP275621

Rabbit Monoclonal Antibody

PKC gamma Monoclonal specifically detects PKC gamma in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen PKC gamma
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone JG69-32
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias EC 2.7.11, EC 2.7.11.13, MGC57564, PKCCSCA14, PKC-gamma, PKCGprotein kinase C gamma type, protein kinase C, gamma
Gene Symbols PRKCG
Host Species Rabbit
Immunogen Recombinant protein within Human PKC gamma aa 564-697/ 697. (SwissProt: P05129 Human; SwissProt: P63318 Mouse; SwissProt: P63319 Rat)
Molecular Weight of Antigen 78 kDa
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Protein Kinase, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5582.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.