Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKC iota Mouse anti-Human, Clone: PRKCI/4912, Novus Biologicals™
SDP

Catalog No. NB177463 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB177463 20 μg
NB177462 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB177463 Supplier Novus Biologicals Supplier No. NBP31375320UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PKC iota Monoclonal antibody specifically detects PKC iota in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen PKC iota
Applications Immunohistochemistry (Paraffin), Peptide Array
Classification Monoclonal
Clone PRKCI/4912
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Formulation 10 mM PBS with 0.05% BSA
Gene Accession No. P41743
Gene Alias aPKC-lambda/iota, Atypical protein kinase C-lambda/iota, DXS1179E, EC 2.7.11, EC 2.7.11.13, MGC26534, nPKC-iota, PKCI, PRKC-lambda/iota, protein kinase C iota type, protein kinase C, iota
Host Species Mouse
Immunogen Recombinant fragment (around aa100-300) of human PKC iota protein (exact sequence is proprietary) (Uniprot: P41743)
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5584
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.