Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKC theta Antibody (453416), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB370278 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB370278 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB370278 Supplier Novus Biologicals Supplier No. FAB4368E
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PKC theta Monoclonal specifically detects PKC theta in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining), Western Blot.

Specifications

Antigen PKC theta
Applications CyTOF, Flow Cytometry (Intracellular Staining), Western Blot
Classification Monoclonal
Clone 453416
Conjugate DyLight 405
Dilution Western Blot, Simple Western, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.11, EC 2.7.11.13, MGC126514, MGC141919, nPKC-theta, PRKCT, protein kinase C theta type, protein kinase C, theta
Host Species Mouse
Immunogen E. coli-derived recombinant human PKC theta , His580-Ser706, Accession # Q04759
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5588
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.