Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PKC theta Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00
Specifications
Antigen | PKC theta |
---|---|
Dilution | Western Blot -Reported in scientific literature (PMID:34073294), Immunocytochemistry/ Immunofluorescence 0.25-2 μg/ml |
Applications | Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
PKC theta Polyclonal antibody specifically detects PKC theta in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
PKC theta | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Protein Kinase, Signal Transduction | |
PBS (pH 7.2) and 40% Glycerol | |
5588 | |
IgG | |
Affinity purified |
Western Blot -Reported in scientific literature (PMID:34073294), Immunocytochemistry/ Immunofluorescence 0.25-2 μg/ml | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse | |
EC 2.7.11, EC 2.7.11.13, MGC126514, MGC141919, nPKC-theta, PRKCT, protein kinase C theta type, protein kinase C, theta | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence: SPFLRIGLSNFDCGSCQSCQGEAVNPYCAVLVKEYVESENGQMYIQK | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title