Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKCbeta Rabbit anti-Human, Clone: ST48-06, Novus Biologicals™
SDP

Catalog No. NBP267516 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267516 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267516 Supplier Novus Biologicals Supplier No. NBP267516

Rabbit Monoclonal Antibody

PKC beta Monoclonal specifically detects PKC beta in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen PKCbeta
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone ST48-06
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias EC 2.7.11, EC 2.7.11.13, MGC41878, PKC-B, PKC-beta, PKCBPRKCB2, PRKCB1protein kinase C beta 1, protein kinase C beta type, protein kinase C, beta, protein kinase C, beta 1, protein kinase C, beta 1 polypeptide
Gene Symbols PRKCB
Host Species Rabbit
Immunogen Synthetic peptide within Human PKC beta aa 615-660/ 671. (SwissProt: P05771 Human; SwissProt: P68404 Mouse; SwissProt: P68403 Rat)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Cancer, mTOR Pathway, Phospho Specific, Protein Kinase, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5579
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.