Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKCmu Rabbit anti-Human, Clone: JM30-17, Novus Biologicals™
SDP

Catalog No. NBP267066 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267066 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267066 Supplier Novus Biologicals Supplier No. NBP267066

Rabbit Monoclonal Antibody

PKC mu Monoclonal specifically detects PKC mu in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen PKCmu
Applications Western Blot
Classification Monoclonal
Clone JM30-17
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias EC 2.7.11, nPKC-D1, nPKC-mu, PKCM, PKC-MU, PKD1, PKDEC 2.7.11.13, PRKCMPKC-mu, Protein kinase C mu type, protein kinase C, mu, Protein kinase D, protein kinase D1, serine/threonine-protein kinase D1
Gene Symbols PRKD1
Host Species Rabbit
Immunogen Synthetic peptide within Human PKC mu aa 201-250/ 912. (SwissProt: Q15139 Human; SwissProt: Q62101 Mouse; SwissProt: Q9WTQ1 Rat)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Golgi Apparatus Markers, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5587
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.